Ethyl 2-amino-2-(3-chloropyridin-4-yl)acetate hydrochloride 97%

Supplier: Ambeed
Warning

A375682-100MG A375682-250MG
77240-320EA 9.27 USD
77240-320 77240-322
Ethyl 2-amino-2-(3-chloropyridin-4-yl)acetate hydrochloride 97%
Ethyl 2-amino-2-(3-chloropyridin-4-yl)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂Cl₂N₂O₂
MW: 251.11 g/mol
MDL Number: MFCD30186146
CAS Number: 1956319-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR