(S,S)-1,1'-Bis(dicyclohexylphosphino)-2,2'-bis[(S)-α-(dimethylamino)benzyl]ferrocene 98%
Supplier: Ambeed
|
Warning
|
A375646-100MG
A375646-1G
A375646-250MG
A375646-50MG
77251-840EA
66
USD
77251-840
77251-842
77251-844
77251-846
(S,S)-1,1'-Bis(dicyclohexylphosphino)-2,2'-bis[(S)-α-(dimethylamino)benzyl]ferrocene 98%
(S,S)-1,1'-Bis(dicyclohexylphosphino)-2,2'-bis[(S)-α-(dimethylamino)benzyl]ferrocene
Precaución: for Research Use Only
Formula:
C₅₂H₇₄FeN₂P₂ |
MDL Number:
MFCD08561143 CAS Number: 849924-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...