5,5'-(Ethyne-1,2-diyl)diisophthalic acid 98%

Supplier: Ambeed
Warning

A375548-1G A375548-250MG A375548-5G
77264-168EA 262.65 USD
77264-168 77264-170 77264-172
5,5'-(Ethyne-1,2-diyl)diisophthalic acid 98%
5,5'-(Ethyne-1,2-diyl)diisophthalic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₀O₈
MW: 354.26 g/mol
CAS Number: 957014-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR