4-Methoxyquinolin-3-amine 97%

Supplier: Ambeed
Warning

A375415-100MG A375415-1G A375415-250MG
77240-314EA 119.48 USD
77240-314 77240-316 77240-318
4-Methoxyquinolin-3-amine 97%
4-Methoxyquinolin-3-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MW: 174.19 g/mol
MDL Number: MFCD12403918
CAS Number: 103040-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR