2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulfonate 97%

Supplier: Ambeed
Warning

A375232-1G A375232-5G
77251-834EA 12 USD
77251-834 77251-836
2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulfonate 97%
2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₁₅H₂₃F₃O₄S
MDL Number: MFCD00229854
CAS Number: 59643-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR