4-(1H-Pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A375023-100MG A375023-250MG
77264-164EA 103 USD
77264-164 77264-166
4-(1H-Pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidine 97%
4-(1H-Pyrazol-4-yl)-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₉H₇N₅
MW: 185.185 g/mol
MDL Number: MFCD20683197
CAS Number: 952518-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR