4-(2-Aminoethoxy)-N2,N6-bis(4-(2-aminoethoxy)quinolin-2-yl)pyridine-2,6-dicarboxamide 2,2,2-trifluoroacetate 98%

Supplier: Ambeed
Warning

A374986-1MG A374986-5MG
77264-160EA 35.02 USD
77264-160 77264-162
4-(2-Aminoethoxy)-N2,N6-bis(4-(2-aminoethoxy)quinolin-2-yl)pyridine-2,6-dicarboxamide 2,2,2-trifluoroacetate 98%
4-(2-Aminoethoxy)-N2,N6-bis(4-(2-aminoethoxy)quinolin-2-yl)pyridine-2,6-dicarboxamide 2,2,2-trifluoroacetate

Precaución: for Research Use Only

Formula: C₃₃H₃₃F₃N₈O₇
MW: 710.65 g/mol
MDL Number: MFCD26142941
CAS Number: 1472611-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR