2-Amino-4,6-dichlorophenol 95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloro-6-hydroxyaniline

A374947-10G A374947-1G A374947-5G A374947-100G
77290-900EA 53.56 USD
77290-900 77290-902 77290-904 77748-750
2-Amino-4,6-dichlorophenol 95%
2-Amino-4,6-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.01 g/mol
MDL Number: MFCD00035766
CAS Number: 527-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR