2-Acetyl-3-bromothiophene 96%

Supplier: Ambeed
Warning

Synonyms: 2-Acetyl-3-boromothiophene, 1-(3-Bromo-2-thienyl)-1-ethanone

A374917-100G A374917-10G A374917-1G A374917-250MG A374917-25G A374917-5G
77251-822EA 621.09 USD
77251-822 77251-824 77251-826 77251-828 77251-830 77251-832
2-Acetyl-3-bromothiophene 96%
2-Acetyl-3-bromothiophene

Precaución: for Research Use Only

Formula: C₆H₅BrOS
MW: 205.08 g/mol
MDL Number: MFCD00794287
CAS Number: 42877-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR