2-Azaspiro[4.4]nonan-3-one 98%

Supplier: Ambeed
Warning

A374846-1G A374846-250MG A374846-5G
77264-154EA 56 USD
77264-154 77264-156 77264-158
2-Azaspiro[4.4]nonan-3-one 98%
2-Azaspiro[4.4]nonan-3-one

Precaución: for Research Use Only

Formula: C₈H₁₃NO
MDL Number: MFCD09864498
CAS Number: 75751-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR