(3-Chlorophenyl)acetic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chlorophenylacetic acid
A374833-1000G
A374833-100G
A374833-10G
A374833-25G
A374833-500G
A374833-5G
77278-476EA
213.21
USD
77278-476
77278-478
77278-480
77278-482
77278-484
77278-486
(3-Chlorophenyl)acetic acid 97%
(3-Chlorophenyl)acetic acid
Precaución: for Research Use Only
Formula:
(C₆H₄Cl)CH₂COOH MW: 170.6 g/mol |
MDL Number:
MFCD00004332 CAS Number: 1878-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...