4-Amino-1,1,1-trifluorobut-3-en-2-one 97%

Supplier: Ambeed
Danger

Synonyms: 4-Amino-2-oxo-1,1,1-trifluorobut-3-ene, 4-Amino-1,1,1-trifluoro-3-buten-2-one

A374722-1G A374722-250MG A374722-5G
77264-148EA 52.53 USD
77264-148 77264-150 77264-152
4-Amino-1,1,1-trifluorobut-3-en-2-one 97%
4-Amino-1,1,1-trifluorobut-3-en-2-one

Precaución: for Research Use Only

Formula: C₄H₄F₃NO
MW: 139.076 g/mol
MDL Number: MFCD01569588
CAS Number: 184848-89-3
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR