6-Carboxyoxindole 98%

Supplier: Ambeed
Warning

Synonyms: 2-Oxoindoline-6-carboxylic acid

A374155-10G A374155-1G A374155-250MG A374155-5G
77278-468EA 177.16 USD
77278-468 77278-470 77278-472 77278-474
6-Carboxyoxindole 98%
6-Carboxyoxindole

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.1600 g/mol
MDL Number: MFCD01415801
CAS Number: 334952-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR