6-(2-Chloro-4-(6-methylpyrazin-2-yl)phenyl)-8-ethyl-2-((2-(1-methylpiperidin-4-yl)ethyl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one 98+%

Supplier: Ambeed
Warning

Synonyms: FRAX1036

A374154-10MG A374154-1MG A374154-25MG A374154-50MG A374154-5MG
77240-294EA 39.14 USD
77240-294 77240-296 77240-298 77240-300 77240-302
6-(2-Chloro-4-(6-methylpyrazin-2-yl)phenyl)-8-ethyl-2-((2-(1-methylpiperidin-4-yl)ethyl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one 98+%
6-(2-Chloro-4-(6-methylpyrazin-2-yl)phenyl)-8-ethyl-2-((2-(1-methylpiperidin-4-yl)ethyl)amino)pyrido[2,3-d]pyrimidin-7(8H)-one

Precaución: for Research Use Only

Formula: C₂₈H₃₂ClN₇O
MW: 518.05 g/mol
MDL Number: MFCD30187513
CAS Number: 1432908-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR