2-Butyne-1-ol 98%

Supplier: Ambeed
Danger

Synonyms: 2-Butyn-1-ol, But-2-yn-1-ol, 1-Hydroxy-2-butyne

A374099-1G A374099-25G A374099-5G
77264-142EA 5 USD
77264-142 77264-144 77264-146
2-Butyne-1-ol 98%
2-Butyne-1-ol

Precaución: for Research Use Only

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 142
Melting Pt: -2
MDL Number: MFCD00002914
CAS Number: 764-01-2
UN: 1987
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR