o-Ethyltoluene 98%

Supplier: Ambeed
Danger

Synonyms: 1-Ethyl-2-methylbenzene, 2-Ethyltoluene

A373861-1G A373861-25G A373861-5G
77264-136EA 5.15 USD
77264-136 77264-138 77264-140
o-Ethyltoluene 98%
o-Ethyltoluene

Precaución: for Research Use Only

Formula: C₉H₁₂
MW: 120.19 g/mol
MDL Number: MFCD00009257
CAS Number: 611-14-3
UN: 3295
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR