4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl 98%

Supplier: Ambeed
Danger

Synonyms: TEMPONE, 4-Oxo-2,2,6,6-tetramethylpiperidinooxy, 4-Oxo-TEMPO

A373622-1G A373622-25G A373622-5G
77290-894EA 6.18 USD
77290-894 77290-896 77290-898
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl 98%
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl

Precaución: for Research Use Only

Formula: C₉H₁₆NO₂
MW: 170.23 g/mol
MDL Number: MFCD00006187
CAS Number: 2896-70-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR