4,4',4''-((1,3,5-Triazine-2,4,6-triyl)tris(oxy))tribenzaldehyde 97%

Supplier: Ambeed
Warning

A373444-100MG A373444-1G A373444-250MG A373444-5G
77264-128EA 26.78 USD
77264-128 77264-130 77264-132 77264-134
4,4',4''-((1,3,5-Triazine-2,4,6-triyl)tris(oxy))tribenzaldehyde 97%
4,4',4''-((1,3,5-Triazine-2,4,6-triyl)tris(oxy))tribenzaldehyde

Precaución: for Research Use Only

Formula: C₂₄H₁₅N₃O₆
MW: 441.39 g/mol
MDL Number: MFCD31700811
CAS Number: 3140-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR