2-(2,5-Dichlorophenyl)-1,4-benzoquinone 98%

Supplier: Ambeed
Warning

Synonyms: 2',5'-Dichloro-[1,1'-biphenyl]-2,5-dione, TPI-1

A373292-10MG A373292-1MG A373292-50MG A373292-5MG
77278-456EA 21.63 USD
77278-456 77278-458 77278-460 77278-462
2-(2,5-Dichlorophenyl)-1,4-benzoquinone 98%
2-(2,5-Dichlorophenyl)-1,4-benzoquinone

Precaución: for Research Use Only

Formula: C₁₂H₆Cl₂O₂
MW: 253.081 g/mol
MDL Number: MFCD00448731
CAS Number: 79756-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR