1-Phenylpyrrole 95%

Supplier: Ambeed
Warning

Synonyms: N-Phenylpyrrole, 1-Phenyl-1H-pyrrole

A373281-100G A373281-10G A373281-1G A373281-25G A373281-5G
77240-282EA 332.69 USD
77240-282 77240-284 77240-286 77240-288 77240-290
1-Phenylpyrrole 95%
1-Phenylpyrrole

Precaución: for Research Use Only

Formula: C₁₀H₉N
MW: 143.19 g/mol
MDL Number: MFCD00005343
CAS Number: 635-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR