Methyl 3-(1,3-dioxolan-2-yl)benzoate 97%
Supplier: Ambeed
|
Warning
|
A372685-100MG
A372685-250MG
77240-278EA
48.41
USD
77240-278
77240-280
Methyl 3-(1,3-dioxolan-2-yl)benzoate 97%
Methyl 3-(1,3-dioxolan-2-yl)benzoate
Precaución: for Research Use Only
Formula:
C₁₁H₁₂O₄ MW: 208.21 g/mol |
MDL Number:
MFCD06209425 CAS Number: 124038-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...