5,6-Difluorooxindole 95%

Supplier: Ambeed
Warning

Synonyms: 5,6-Difluoroindolin-2-one

A372364-10G A372364-1G A372364-250MG A372364-5G
77278-438EA 441.87 USD
77278-438 77278-440 77278-442 77278-444
5,6-Difluorooxindole 95%
5,6-Difluorooxindole

Precaución: for Research Use Only

Formula: C₈H₅F₂NO
MW: 169.13 g/mol
MDL Number: MFCD01075216
CAS Number: 71294-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR