4-Hydroxy-1-naphthaldehyde 95%

Supplier: Ambeed
Warning

A372363-100MG A372363-1G A372363-250MG A372363-25G
77290-878EA 13.39 USD
77290-878 77290-880 77290-882 77290-884
4-Hydroxy-1-naphthaldehyde 95%
4-Hydroxy-1-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈O₂
MW: 172.18 g/mol
MDL Number: MFCD00053486
CAS Number: 7770-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR