3,6,9,12-Tetraoxapentadec-14-yn-1-ol 96%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG4-alcohol

A372250-1G A372250-250MG A372250-5G
77264-108EA 118.45 USD
77264-108 77264-110 77264-112
3,6,9,12-Tetraoxapentadec-14-yn-1-ol 96%
3,6,9,12-Tetraoxapentadec-14-yn-1-ol

Precaución: for Research Use Only

Formula: C₁₁H₂₀O₅
MW: 232.27 g/mol
MDL Number: MFCD22201537
CAS Number: 87450-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR