2-Fluoro-3-(trifluoromethyl)benzamide 95+%

Supplier: Ambeed
Warning

A372012-1G A372012-25G A372012-5G
77240-266EA 5 USD
77240-266 77240-268 77240-270
2-Fluoro-3-(trifluoromethyl)benzamide 95+%
2-Fluoro-3-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₈H₅F₄NO
MDL Number: MFCD00061146
CAS Number: 207853-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR