(2-Bromo-4-(trifluoromethyl)phenyl)methanamine 95%

Supplier: Ambeed
Warning

A371840-100MG A371840-1G A371840-250MG
77240-260EA 37.08 USD
77240-260 77240-262 77240-264
(2-Bromo-4-(trifluoromethyl)phenyl)methanamine 95%
(2-Bromo-4-(trifluoromethyl)phenyl)methanamine

Precaución: for Research Use Only

Formula: C₈H₇BrF₃N
MW: 254.04 g/mol
MDL Number: MFCD13185400
CAS Number: 1001109-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR