1-(Bromomethyl)-3-chloro-2-methylbenzene 95%

Supplier: Ambeed
Warning

A371206-1G A371206-250MG A371206-5G
77264-102EA 99 USD
77264-102 77264-104 77264-106
1-(Bromomethyl)-3-chloro-2-methylbenzene 95%
1-(Bromomethyl)-3-chloro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrCl
MW: 219.50 g/mol
MDL Number: MFCD12025000
CAS Number: 90369-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR