2-Chloro-N-methyl-N-phenylacetamide 98%

Supplier: Ambeed
Danger

A370925-1G A370925-250MG
77264-092EA 19 USD
77264-092 77264-094
2-Chloro-N-methyl-N-phenylacetamide 98%
2-Chloro-N-methyl-N-phenylacetamide

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO
MDL Number: MFCD00028184
CAS Number: 2620-05-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR