3-(Methylamino)propionitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3-(N-Methylamino)propionitrile, N-(2-Cyanoethyl)methylamine, N-Methyl-β-alaninenitrile, 3-(Methylamino)propanenitrile

A370808-100G A370808-500G
77278-430EA 64.89 USD
77278-430 77748-740
3-(Methylamino)propionitrile 97%
3-(Methylamino)propionitrile

Precaución: for Research Use Only

Formula: C₄H₈N₂
MW: 84.12 g/mol
MDL Number: MFCD00001954
CAS Number: 693-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR