ATN-161 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-((R)-2-((S)-2-((S)-2-((S)-1-Acetylpyrrolidine-2-carboxamido)-3-(1H-imidazol-4-yl)propanamido)-3-hydroxypropanamido)-3-mercaptopropanamido)succinamide

A370715-10MG A370715-1MG A370715-50MG A370715-5MG
77240-246EA 21.63 USD
77240-246 77240-248 77240-250 77240-252
ATN-161 98%
ATN-161

Precaución: for Research Use Only

Formula: C₂₃H₃₅N₉O₈S
MW: 597.64 g/mol
MDL Number: MFCD30478885
CAS Number: 262438-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR