2-(1-Adamantyl)-4-bromoanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Bromo-2-methoxyphenyl)adamantane, 1-(5-Bromo-2-methoxy-phenyl)adamantane

A370703-100G A370703-25G A370703-500G A370703-5G
77290-868EA 135 USD
77290-868 77290-870 77290-872 77290-874
2-(1-Adamantyl)-4-bromoanisole 98%
2-(1-Adamantyl)-4-bromoanisole

Precaución: for Research Use Only

Formula: C₁₇H₂₁BrO
MDL Number: MFCD03855308
CAS Number: 104224-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR