4-((11-(Acryloyloxy)undecyl)oxy)benzoic acid 97%

Supplier: Ambeed
Warning

A370699-10G A370699-25G A370699-5G
77240-242EA 194.67 USD
77240-242 77240-244 77746-749
4-((11-(Acryloyloxy)undecyl)oxy)benzoic acid 97%
4-((11-(Acryloyloxy)undecyl)oxy)benzoic acid

Precaución: for Research Use Only

Formula: C₂₁H₃₀O₅
MW: 362.45 g/mol
MDL Number: MFCD23098832
CAS Number: 106620-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR