trans-4-(4-Propylcyclohexyl)benzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-(trans-4-propylcyclohexyl)benzonitrile

A370500-100G A370500-10G A370500-1G A370500-25G A370500-5G
77278-420EA 114 USD
77278-420 77278-422 77278-424 77278-426 77278-428
trans-4-(4-Propylcyclohexyl)benzonitrile 98%
trans-4-(4-Propylcyclohexyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₆H₂₁N
MDL Number: MFCD04038799
CAS Number: 61203-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR