2,3-Dihydrobenzo[b]furan 98%

Supplier: Ambeed
Warning

A370289-100G A370289-10G A370289-25G A370289-500G A370289-5G
77251-754EA 47.38 USD
77251-754 77251-756 77251-758 77251-760 77251-762
2,3-Dihydrobenzo[b]furan 98%
2,3-Dihydrobenzo[b]furan

Precaución: for Research Use Only

Formula: C₈H₈O
MW: 120.15 g/mol
MDL Number: MFCD00005855
CAS Number: 496-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR