XMD17-109 98%

Supplier: Ambeed
Warning

Synonyms: XMD17109, XMD17-109, 11-cyclopentyl-2-[2-ethoxy-4-[4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl]anilino]-5-methylpyrimido[4,5-b][1,4]benzodiazepin-6-one, XMD17 109, 11-Cyclopentyl-2-((2-ethoxy-4-(4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl)phenyl)amino)-5-methyl-5,11-dihydro-6H-benzo[e]pyrimido[5,4-b][1,4]diazepin-6-one

A370232-10MG A370232-1MG A370232-2MG A370232-50MG A370232-5MG
77240-232EA 113.3 USD
77240-232 77240-234 77240-236 77240-238 77240-240
XMD17-109 98%
XMD17-109

Precaución: for Research Use Only

Formula: C₃₆H₄₆N₈O₃
MW: 638.8 g/mol
MDL Number: MFCD26142929
CAS Number: 1435488-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR