5-(Aminomethyl)-2-fluorophenol hydrochloride 97%

Supplier: Ambeed
Warning

A369993-1G A369993-250MG
77251-750EA 217.33 USD
77251-750 77251-752
5-(Aminomethyl)-2-fluorophenol hydrochloride 97%
5-(Aminomethyl)-2-fluorophenol hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉ClFNO
MW: 177.60 g/mol
CAS Number: 2044704-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR