1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-2′-O-methyl-uridine

A369665-1G A369665-250MG A369665-5G
77278-410EA 49.44 USD
77278-410 77278-412 77278-414
1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione 97%
1-((2R,3R,4R,5R)-4-Hydroxy-5-(hydroxymethyl)-3-methoxytetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₆
MW: 272.25 g/mol
MDL Number: MFCD03788700
CAS Number: 55486-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR