3,5,5-Trimethylhexyl acrylate 90% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A369571-5G A369571-1G
77251-746EA 69 USD
77251-746 77746-744
3,5,5-Trimethylhexyl acrylate 90% stabilized
3,5,5-Trimethylhexyl acrylate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₂
MDL Number: MFCD00080661
CAS Number: 45125-03-9
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR