3-Hydroxy-4-methoxy-2-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A368645-100MG A368645-250MG
77278-406EA 40.17 USD
77278-406 77278-408
3-Hydroxy-4-methoxy-2-nitrobenzaldehyde 97%
3-Hydroxy-4-methoxy-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₅
MW: 197.15 g/mol
MDL Number: MFCD00031118
CAS Number: 6284-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR