6-Chloronicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloropyridine-3-carboxylic acid, 6-Chloro-3-pyridinecarboxylic acid

A367625-1000G A367625-100G A367625-10G A367625-25G A367625-500G A367625-5G
77278-388EA 213.21 USD
77278-388 77278-390 77278-392 77278-394 77278-396 77278-398
6-Chloronicotinic acid 98%
6-Chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
MDL Number: MFCD00006241
CAS Number: 5326-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR