2-Amino-2-(hydroxymethyl)propane-1,3-diol (3-((4-((2-amino-3-chloropyridin-4-yl)oxy)-3-fluorophenyl)carbamoyl)-5-(4-fluorophenyl)-4-oxopyridin-1(4H)-yl)methyl phosphate 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
SCR-1481B1
A366964-10MG
A366964-1MG
A366964-25MG
A366964-5MG
77240-202EA
77.25
USD
77240-202
77240-204
77240-206
77240-208
2-Amino-2-(hydroxymethyl)propane-1,3-diol (3-((4-((2-amino-3-chloropyridin-4-yl)oxy)-3-fluorophenyl)carbamoyl)-5-(4-fluorophenyl)-4-oxopyridin-1(4H)-yl)methyl phosphate 98+%
2-Amino-2-(hydroxymethyl)propane-1,3-diol (3-((4-((2-amino-3-chloropyridin-4-yl)oxy)-3-fluorophenyl)carbamoyl)-5-(4-fluorophenyl)-4-oxopyridin-1(4H)-yl)methyl phosphate
Precaución: for Research Use Only
Formula:
C₂₈H₂₉ClF₂N₅O₁₀P MW: 699.98 g/mol |
MDL Number:
MFCD30187512 CAS Number: 1174161-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...