Methyltriphenylsilane 98%

Supplier: Ambeed
Warning

A366287-100G A366287-1G A366287-25G A366287-5G
77251-710EA 146 USD
77251-710 77251-712 77251-714 77251-716
Methyltriphenylsilane 98%
Methyltriphenylsilane

Precaución: for Research Use Only

Formula: C₁₉H₁₈Si
MW: 274.44 g/mol
MDL Number: MFCD00041325
CAS Number: 791-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR