4-Bromo-6-methoxyquinoline 95%

Supplier: Ambeed
Warning

A366161-100MG A366161-10G A366161-1G A366161-250MG A366161-5G
77290-846EA 23.69 USD
77290-846 77290-848 77290-850 77290-852 77290-854
4-Bromo-6-methoxyquinoline 95%
4-Bromo-6-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MW: 238.0800 g/mol
MDL Number: MFCD08437074
CAS Number: 42881-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR