3-Chloro-5-fluorobenzyl alcohol 98%

Supplier: Ambeed
Warning

A365787-1G A365787-25G A365787-5G
77278-382EA 16.48 USD
77278-382 77278-384 77278-386
3-Chloro-5-fluorobenzyl alcohol 98%
3-Chloro-5-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆ClFO
MW: 160.58 g/mol
MDL Number: MFCD03788554
CAS Number: 79944-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR