4-Butoxybenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (4-Butoxyphenyl)methanol

A365775-10G A365775-5G
77290-842EA 58.71 USD
77290-842 77290-844
4-Butoxybenzyl alcohol 98%
4-Butoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
MW: 180.25 g/mol
MDL Number: MFCD00004657
CAS Number: 6214-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR