(R)-(+)-2,2'-Bis(di-i-propylphosphino)-6,6'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: Ambeed
Warning

A365583-250MG A365583-100MG
77251-706EA 281.99 USD
77251-706 77251-708 77746-729
(R)-(+)-2,2'-Bis(di-i-propylphosphino)-6,6'-dimethoxy-1,1'-biphenyl ≥97%
(R)-(+)-2,2'-Bis(di-i-propylphosphino)-6,6'-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₂₆H₄₀O₂P₂
MDL Number: MFCD09753011
CAS Number: 150971-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR