2-Benzyloxy-1,3-propanediol 98%

Supplier: Ambeed
Warning

A365428-250MG A365428-1G A365428-25G
77264-060EA 15 USD
77264-060 77616-596 77746-728
2-Benzyloxy-1,3-propanediol 98%
2-Benzyloxy-1,3-propanediol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MDL Number: MFCD00075493
CAS Number: 14690-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR