Arachidic acid 98% (by HPLC)
Supplier: Ambeed
|
Warning
|
A365161-100G
A365161-1G
A365161-25G
A365161-5G
77290-828EA
305
USD
77290-828
77290-830
77290-832
77594-086
Arachidic acid 98% (by HPLC)
Arachidic acid
Precaución: for Research Use Only
Formula:
C₂₀H₄₀O₂ |
MDL Number:
MFCD00002755 CAS Number: 506-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...