(R)-3,3'-Bis(9-phenanthrenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthyl HydrogenPhosphate 98%

Supplier: Ambeed
Warning

A365061-100MG A365061-1G A365061-250MG
77251-700EA 119.48 USD
77251-700 77251-702 77251-704
(R)-3,3'-Bis(9-phenanthrenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthyl HydrogenPhosphate 98%
(R)-3,3'-Bis(9-phenanthrenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthyl HydrogenPhosphate

Precaución: for Research Use Only

Formula: C₄₈H₃₇O₄P
MW: 708.77 g/mol
MDL Number: MFCD31560049
CAS Number: 934201-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR