4-(Trifluoromethylthio)phenylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-((Trifluoromethyl)thio)phenyl)acetic acid, 4-(Carboxymethyl)phenyl trifluoromethyl sulphide, {4-[(Trifluoromethyl)sulphanyl]phenyl}acetic acid, 4-[(Trifluoromethyl)sulphanyl]phenylacetic acid

A364908-100G A364908-1G A364908-25G A364908-5G
77264-052EA 891.01 USD
77264-052 77264-054 77264-056 77264-058
4-(Trifluoromethylthio)phenylacetic acid 98%
4-(Trifluoromethylthio)phenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂S
MDL Number: MFCD00236351
CAS Number: 243977-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR